Please login to your online account to display your discounted pricing

anti-NFkB p65 NLS, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of the NFkB p65 NLS nuclear localization signal (NLS) (amino acids DTDDRHRIEEKRKRKT) was used as the immunogen for this antibody.
Antigen NFkB p65 NLS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224541

 
novus biologicals canada
NBP224541
Unlabeled 0.1mg Each for $307.00

NBP224541SS

 
novus biologicals canada
NBP224541SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of the NFkB p65 NLS nuclear localization signal (NLS) (amino acids DTDDRHRIEEKRKRKT) was used as the immunogen for this antibody.
Antigen NFkB p65 NLS
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in human, mouse, rat, bull, chicken, chimp, Drosophila, dog, rhesus monkey, Xenopus and zebrafish. It is also 100% homologous with MEK2, with the phospho-serines at positions 222 and 226.
Gene ID (Entrez) 5970
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL