Please login to your online account to display your discounted pricing

anti-NFATC1/NFAT2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056732SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056732T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4772
Gene Accession No. NP_006153.2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Immunogen Amino acids 264-282 (NKRKYSLNGRQPPYSPHHS) of human NFATc1 were used as immunogen for this antibody.
Quantity 0.025mg
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Primary or Secondary Primary
Test Specificity The amino acid sequence used is 90% homologous in mouse and rat and 100% identical in human NFATc1 A, B, C and E isoforms. The reported amino acid size of NFATc1 isoforms are as follows: A -716 aa, B-825 aa, C-930 aa, and E-812 aa.
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Antigen NFATC1/NFAT2
Regulatory Status RUO
Host Species Rabbit