Please login to your online account to display your discounted pricing

anti-NFAM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224943SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224943SS

  • / Each
 

Description & Specifications

Specifications

Antigen NFAM1
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation 100μL of crude rabbit sera with 50% glycerol
Gene Accession No. Q8NET5
Host Species Rabbit
Immunogen Within the range of amino acids 250-278 of human NFAM1 were used as the immunogen.
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 150372
Target Species Human