Please login to your online account to display your discounted pricing

anti-NFAM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224943

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224943

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation 100μL of crude rabbit sera with 50% glycerol
Cross Reactivity Human
Gene Accession No. Q8NET5
Gene ID (Entrez) 150372
Host Species Rabbit
Conjugate Unlabeled
Primary or Secondary Primary
Applications Western Blotting
Antigen NFAM1
Format Antiserum
Regulatory Status RUO
Quantity 0.1mL
Immunogen Within the range of amino acids 250-278 of human NFAM1 were used as the immunogen.