Please login to your online account to display your discounted pricing

anti-NFAM1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 250-278 of human NFAM1 were used as the immunogen.
Antigen NFAM1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224943

 
novus biologicals canada
NBP224943
Unlabeled 0.1mL Each for $259.02

NBP224943SS

 
novus biologicals canada
NBP224943SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 250-278 of human NFAM1 were used as the immunogen.
Antigen NFAM1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. Q8NET5
Gene ID (Entrez) 150372
Formulation 100μL of crude rabbit sera with 50% glycerol