Please login to your online account to display your discounted pricing

anti-Neurotrophin-4 Polyclonal, Thermo Scientific™

Neurotrophin-4 Polyclonal Ab PA1-18392 from Thermo Scientific™ detects Neurotrophin-4 in Hu, Ms and Nhp samples, and is validated for ELISA, IHC, Inhibition Assays and WB

$494.55 - $509.25

Specifications

Antigen Neurotrophin-4
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-human primate
Format Serum
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-644

 
pierce biotechnology inc
PA118392
Unlabeled 100μL Each for $494.55

01-670-645

 
pierce biotechnology inc
PA118393
Unlabeled 500μg Each for $509.25
Description & Specifications

Specifications

Antigen Neurotrophin-4
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-human primate
Format Serum
Host Species Sheep
Immunogen Recombinant human NT4.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Neurotrophin-4 Polyclonal Ab PA1-18392 from Thermo Scientific™ detects Neurotrophin-4 in Hu, Ms and Nhp samples, and is validated for ELISA, IHC, Inhibition Assays and WB

This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
Host Species: Sheep
Cross Reactivity: Human, Mouse, Non-human primate
Isotype: IgG
Immunogen: Recombinant human NT4.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Neurotrophin-5, NT-5, Neutrophic factor 5, Neurotrophin-4, NT-4, Neutrophic factor 4, NTF5, NTF4
Uniprot Accession: P34130, Q80VU4
Gene Symbol: NTF4
Entrez GeneID: 4909, 78405
PA1-18392 detects Neurotrophin-4 in Human, Mouse and Non-human primate samples. PA1-18392 has been successfully used in ELISA, Immunohistochemistry, Inhibition Assays and Western Blot procedures. PA1-18392 The immunogen is Recombinant human NT4..