missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Neurotensin Receptor 2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Neurotensin receptor 2 was used as the immunogen.
Antigen Neurotensin Receptor 2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056472T Novus Biologicals Canada
NB10056472SS
0.025mg Each for $139.00
NB10056472 Novus Biologicals Canada
NB10056472
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
Neurotensin Receptor 2
Affinity Purified
RUO
Reacts with Human and Mouse. Predicted Reactivity: Rat, Dog, Bovine and Mouse.
PBS containing 0.2% gelatin
A portion of amino acids 150-200 of human Neurotensin receptor 2 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
23620
0.5mg/mL