Please login to your online account to display your discounted pricing

anti-Neuropilin-1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 813-839 of rat Neuropilin 1 was used as the immunogen.
Antigen Neuropilin-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224617SS

 
novus biologicals canada
NBP224617SS
Unlabeled 0.025mg Each for $109.00

NBP224617

 
novus biologicals canada
NBP224617
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 813-839 of rat Neuropilin 1 was used as the immunogen.
Antigen Neuropilin-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human. This product is also predicted to cross react with Chimpanzee, Rhesus Monkey, and Orangutan.
Gene Accession No. Q9QWJ9
Gene ID (Entrez) 8829
Formulation 100μg of antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL