Please login to your online account to display your discounted pricing

anti-Neuron Specific Enolase Polyclonal, Thermo Scientific™

Neuron Specific Enolase Polyclonal Ab PA1-10010 from Thermo Scientific™ specifically detects Neuron Specific Enolase in Hu, Ms and Rt samples, and it is validated for IF and WB

$522.90 - $522.90

Specifications

Antigen Neuron Specific Enolase
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-267

 
pierce biotechnology inc
PA110010
Unlabeled 100μL Each for $522.90
Description & Specifications

Specifications

Antigen Neuron Specific Enolase
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Chicken
Immunogen Purified, recombinant human NSE.
Isotype IgY
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Neuron Specific Enolase Polyclonal Ab PA1-10010 from Thermo Scientific™ specifically detects Neuron Specific Enolase in Hu, Ms and Rt samples, and it is validated for IF and WB

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme, a homodimer, is found in mature neurons and cells of neuronal origin. A switch from alpha enolase to gamma enolase occurs in neural tissue during development in rats and primates.
Host Species: Chicken
Cross Reactivity: Human, Mouse, Rat
Isotype: IgY
Immunogen: Purified, recombinant human NSE.
Formulation: 100μL purified Antibody in PBS, with 10mM sodium azide;
Synonyms: Neuron Specific Enolase
Uniprot Accession: P07323, P09104, P17183
Gene Symbol: ENO2
Entrez GeneID: 13807, 2026, 24334
PA1-10010 detects Neuron Specific Enolase in Human, Mouse and Rat samples. PA1-10010 has been successfully used in Immunofluorescence and Western Blot procedures. PA1-10010 The immunogen is Purified, recombinant human NSE..