Neuroligin 3 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB15514
Catalog No. AB15514MI
Description
Description
This Anti-Neuroligin 3 Antibody is validated for use in western blot, immunohistochemistry, ELISA for the detection of Neuroligin 3.
Specifications
Specifications
Neuroligin 3
Polyclonal
Liquid in PBS with 0.1% BSA and 0.05% sodium azide
Rabbit
50 μg
NM_018977.2
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
ELISA, Immunohistochemistry, Western Blot
Unconjugated
NLGN3; HNL3; NL3; AUTSX1; ASPGX1; KIAA1480
Synthetic peptide near the C-terminus of human Neuroligin 3
Primary
Human
Liquid