Please login to your online account to display your discounted pricing

anti-Neurokinin B, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of mouse proNeurokinin B (∽93% homology to the rat sequence). [UniProt No. P55099]
Antigen Neurokinin B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300201SS

 
novus biologicals canada
NB300201SS
Unlabeled 0.025mL Each for $144.63

NB300201

 
novus biologicals canada
NB300201
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of mouse proNeurokinin B (∽93% homology to the rat sequence). [UniProt No. P55099]
Antigen Neurokinin B
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 6866
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL