Please login to your online account to display your discounted pricing

anti-Neurofilament, Medium chain Polyclonal, Thermo Scientific™

Neurofilament, Medium chain Polyclonal Ab PA1-10001 detects Neurofilament, Medium chain in Bv, Ck, Eq, Hu, Ms, Po and Rt samples, and is validated for ICC, IF, IHC and WB

$480.90 - $522.90

Specifications

Antigen Neurofilament, Medium chain
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-258

 
pierce biotechnology inc
PA110001
Unlabeled 100μL Each for $480.90

01-670-272

 
pierce biotechnology inc
PA110015
Unlabeled 100μL Each for $522.90
Description & Specifications

Specifications

Antigen Neurofilament, Medium chain
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Host Species Chicken
Immunogen Partial recombinant protein corresponding to the C-terminus of NF-M
Isotype IgY
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Neurofilament, Medium chain Polyclonal Ab PA1-10001 detects Neurofilament, Medium chain in Bv, Ck, Eq, Hu, Ms, Po and Rt samples, and is validated for ICC, IF, IHC and WB

Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. The neurofilament (NF) triplet proteins (∽70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are normally restricted to neurons. The 70 kDa NF-protein can self-assemble into a filamentous structure, whereas the 160 kDa and 200 kDa NF-proteins require the presence of the 70 kDa NF-protein to co-assemble.
Host Species: Chicken
Cross Reactivity: Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat
Isotype: IgY
Immunogen: Partial recombinant protein corresponding to the C-terminus of NF-M
Formulation: 100μL purified Antibody in PBS, with 10mM sodium azide;
Synonyms: NFM, NF-M, NMC,160 kD neurofilament protein, neurofilament medium polypeptide, neurofilament 3, NEF3
Uniprot Accession: O77788, P07197, P08552, P08553, P12839, P16053
Gene Symbol: NEFM
Entrez GeneID: 100054178, 18040, 24588, 281347, 396206, 4741
PA1-10001 detects Neurofilament, Medium chain in Bovine, Chicken, Equine, Human, Mouse, Porcine and Rat samples. PA1-10001 has been successfully used in Immunocytochemistry, Immunofluorescence, Immunohistochemistry and Western Blot procedures. PA1-10001 The immunogen is Partial recombinant protein corresponding to the C-terminus of NF-M.