Please login to your online account to display your discounted pricing

anti-Neurofilament, Heavy chain Polyclonal, Thermo Scientific™

Neurofilament, Heavy chain Polyclonal Ab PA1-10016 from Thermo Scientific™ detects Neurofilament, Heavy chain in Bv and Rt samples, and is validated for ICC, IF, IHC and WB

$448.05 - $448.05

Specifications

Antigen Neurofilament, Heavy chain
Cross Reactivity Bovine
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-273

 
pierce biotechnology inc
PA110016
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen Neurofilament, Heavy chain
Cross Reactivity Bovine
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Native NF-H purified from bovine spinal cord
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Neurofilament, Heavy chain Polyclonal Ab PA1-10016 from Thermo Scientific™ detects Neurofilament, Heavy chain in Bv and Rt samples, and is validated for ICC, IF, IHC and WB

Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. The neurofilament (NF) triplet proteins (∽70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are normally restricted to neurons. The 70 kDa NF-protein can self-assemble into a filamentous structure, whereas the 160 kDa and 200 kDa NF-proteins require the presence of the 70 kDa NF-protein to co-assemble.
Host Species: Rabbit
Cross Reactivity: Bovine, Rat
Isotype: IgG
Immunogen: Native NF-H purified from bovine spinal cord
Formulation: 100μL unpurified Antibody in whole serum, with 10mM sodium azide;
Synonyms: NFH, NHC, NF 200 kD
Uniprot Accession: P16884
Gene Symbol: Nefh
Entrez GeneID: 24587, 528842
PA1-10016 detects Neurofilament, Heavy chain in Bovine and Rat samples. PA1-10016 has been successfully used in Immunocytochemistry, Immunofluorescence, Immunohistochemistry and Western Blot procedures. PA1-10016 The immunogen is Native NF-H purified from bovine spinal cord.