missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
NeuN Mouse anti-Rat, Mouse, Cy3, Clone: A60, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB377C3
Description
Detect NeuN using this Anti-NeuN Antibody, clone A60, Cy3 Conjugate validated for use in immunocytochemistry.Specifications
NeuN | |
Monoclonal | |
Cyanine3 | |
NeuN | |
Purified cell nuclei from mouse brain. | |
100 μL | |
Mouse, Rat | |
Purified |
Immunocytochemistry | |
A60 | |
Purified mouse monoclonal IgG1 conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/mL BSA. | |
Mouse | |
Protein A purified | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. Protect from Light. | |
IgG1 |
For Research Use Only