Please login to your online account to display your discounted pricing

anti-Nestin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224391SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224391SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (CRA_a, CRA_b).
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Host Species Rabbit
Gene ID (Entrez) 10763
Conjugate Unlabeled
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 1450-1500 of human Nestin was used as the immunogen.
Quantity 0.025mg
Antigen Nestin
Regulatory Status RUO