Please login to your online account to display your discounted pricing

anti-Nestin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224391

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224391

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (CRA_a, CRA_b).
Quantity 0.1mg
Concentration 0.5mg/mL
Antigen Nestin
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 10763
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Primary or Secondary Primary
Immunogen A portion of amino acids 1450-1500 of human Nestin was used as the immunogen.
Format Purified
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO