Please login to your online account to display your discounted pricing

anti-Nestin, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1450-1500 of human Nestin was used as the immunogen.
Antigen Nestin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224391SS

 
novus biologicals canada
NBP224391SS
Unlabeled 0.025mg Each for $139.00

NBP224391

 
novus biologicals canada
NBP224391
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1450-1500 of human Nestin was used as the immunogen.
Antigen Nestin
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (CRA_a, CRA_b).
Gene ID (Entrez) 10763
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL