Please login to your online account to display your discounted pricing

anti-NEDD4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227201SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227201SS

  • / Each
 

Description & Specifications

Specifications

Antigen NEDD4
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Gene Accession No. P46934
Host Species Rabbit
Immunogen A portion of amino acids 200-260 of human NEDD4 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4734
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, mouse and rat, 94% homologous in zebrafish, and 88% homologous in chicken and opossum.
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat