missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nectin-4/PVRL4 Mouse anti-Human, Clone: N4.61.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT64
Description
Anti-Nectin-4/PVRL4 Antibody, clone N4.61 is an antibody against Nectin-4/PVRL4 for use in immunohistochemistry, ELISA and immunoprecipitation.Specifications
Nectin-4/PVRL4 | |
Monoclonal | |
Unconjugated | |
NP_112178 | |
Mouse | |
Protein G Purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunoprecipitation | |
N4.61.2 | |
Purified mouse monoclonal IgG1κ Supernatant in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PVRL4, PRR4, LNIR | |
Recombinant protein corresponding to human Nectin-4. | |
100 μg | |
Human | |
Purified |
For Research Use Only