Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224478SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224478SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
Antigen NDRG1
Gene ID (Entrez) 10397
Concentration 0.5mg/mL
Format Affinity Purified
Quantity 0.025mg
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Applications Western Blotting
Immunogen A portion of amino acids 350-400 of human NDRG1 was used as the immunogen.
Host Species Goat