Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224478

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224478

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10397
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
Cross Reactivity Human
Regulatory Status RUO
Host Species Goat
Immunogen A portion of amino acids 350-400 of human NDRG1 was used as the immunogen.
Applications Western Blotting
Antigen NDRG1
Quantity 0.1mg
Format Affinity Purified