Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350-400 of human NDRG1 was used as the immunogen.
Antigen NDRG1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223658

 
novus biologicals canada
NBP223658
Unlabeled 0.1mL Each for $255.80

NBP223658SS

 
novus biologicals canada
NBP223658SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 350-400 of human NDRG1 was used as the immunogen.
Antigen NDRG1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_c), dog, rat, monkey and mouse, and 82% homologous in opossum and cow.
Gene ID (Entrez) 10397
Formulation 100μL of crude rabbit sera with 50% glycerol