Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223658SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223658SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL of crude rabbit sera with 50% glycerol
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_c), dog, rat, monkey and mouse, and 82% homologous in opossum and cow.
Monoclonal or Polyclonal Polyclonal
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Gene ID (Entrez) 10397
Quantity 0.025mL
Antigen NDRG1
Regulatory Status RUO
Immunogen A portion of amino acid 350-400 of human NDRG1 was used as the immunogen.
Format Antiserum
Host Species Rabbit