Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223658

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223658

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a and CRA_c), dog, rat, monkey and mouse, and 82% homologous in opossum and cow.
Conjugate Unlabeled
Applications Western Blotting
Regulatory Status RUO
Antigen NDRG1
Gene ID (Entrez) 10397
Cross Reactivity Human
Formulation 100μL of crude rabbit sera with 50% glycerol
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL
Immunogen A portion of amino acid 350-400 of human NDRG1 was used as the immunogen.
Format Antiserum
Host Species Rabbit