missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NDRG1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Goat
Immunogen A portion of amino acids 350-400 of human NDRG1 was used as the immunogen.
Antigen NDRG1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224478 Novus Biologicals Canada
NBP224478
0.1mg Each for $255.80
NBP224478SS Novus Biologicals Canada
NBP224478SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Goat
NDRG1
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and 94% homologous in mouse.
100μg in 200μL PBS containing 0.05% BSA
A portion of amino acids 350-400 of human NDRG1 was used as the immunogen.
Polyclonal
Primary
Western Blot
10397
0.5mg/mL