Please login to your online account to display your discounted pricing

anti-NDM-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177688

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177688

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen NDM-1
Formulation PBS, 30% glycerol
Immunogen A synthetic peptide made to a C-terminal region of the Escherichia coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Regulatory Status RUO
Concentration 1mg/mL
Conjugate Unlabeled
Host Species Rabbit
Applications Western Blotting
Quantity 0.1mL
Cross Reactivity Bacteria