Please login to your online account to display your discounted pricing

anti-NDM-1, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the Escherichia coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Antigen NDM-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177688

 
novus biologicals canada
NBP177688
Unlabeled 0.1mL Each for $329.00

NBP177688SS

 
novus biologicals canada
NBP177688SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the Escherichia coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Antigen NDM-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Formulation PBS, 30% glycerol
Concentration 1mg/mL