missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, PE, Clone: 809220, R&D Systems

$188.76 - $353.45

Specifications

Antigen NCAM-1/CD56
Clone 809220
Format Purified
Host Species Rat
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FAB7820P100 R & D Systems
RDSFAB7820P100
100 Tests Each for $353.45
FAB7820P025 R & D Systems
RDSFAB7820P025
25 Tests Each for $188.76
Specifications

Description

Applications:

Flow Cytometry

Description & Specifications

Specifications

NCAM-1/CD56
Purified
RUO
IgG2a
Primary
809220
Rat
Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56. Leu20-Thr711
Monoclonal
Protein A or G purified from hybridoma culture supernatant