Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, PE, Clone: 301040, R&D Systems


Manufacturer: r & d systems  RDSFAB2408P100

 View more versions of this product

Catalog No. FAB2408P100

  • / Each
 

Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone 301040
Conjugate PE
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56.
Isotype IgG2b
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human