Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, APC, Clone: 809220, R&D Systems


Manufacturer: r & d systems  RDSFAB7820A100

 View more versions of this product

Catalog No. FAB7820A100

  • / Each
 

Description & Specifications

Specifications

Quantity 100 Tests
Host Species Rat
Antigen NCAM-1/CD56
Isotype IgG2a
Cross Reactivity Mouse
Format Purified
Clone 809220
Conjugate APC
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56. Leu20-Thr711
Regulatory Status RUO
Purification Method Protein A or G purified from hybridoma culture supernatant