Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, APC, Clone: 809220, R&D Systems

$151.85 - $419.81

Specifications

Antigen NCAM-1/CD56
Clone 809220
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB7820A025

 
r & d systems
RDSFAB7820A025
APC 25 Tests Each for $151.85

FAB7820A100

 
r & d systems
RDSFAB7820A100
APC 100 Tests Each for $419.81
Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone 809220
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56. Leu20-Thr711
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry