Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, Alexa Fluor 488, Clone: 809220, R&D Systems


Manufacturer: r & d systems  RDSFAB7820G025

 View more versions of this product

Catalog No. FAB7820G025

  • / Each
 

Description & Specifications

Specifications

Clone 809220
Format Purified
Isotype IgG2a
Quantity 25 Tests
Antigen NCAM-1/CD56
Regulatory Status RUO
Conjugate Alexa Fluor 488
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56. Leu20-Thr711
Host Species Rat
Purification Method Protein A or G purified from hybridoma culture supernatant