Please login to your online account to display your discounted pricing

anti-NCAM-1/CD56, Alexa Fluor 488, Clone: 809220, R&D Systems

$129.05 - $363.69

Specifications

Antigen NCAM-1/CD56
Clone 809220
Cross Reactivity Mouse
Format Purified
Host Species Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB7820G025

 
r & d systems
RDSFAB7820G025
Alexa Fluor 488 25 Tests Each for $129.05

FAB7820G100

 
r & d systems
RDSFAB7820G100
Alexa Fluor 488 100 Tests Each for $363.69
Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone 809220
Cross Reactivity Mouse
Format Purified
Host Species Rat
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56. Leu20-Thr711
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry