Please login to your online account to display your discounted pricing

anti-Nbs1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP106705SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP106705SS

  • / Each
 

Description & Specifications

Specifications

Antigen Nbs1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.48mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 150-200). [Swiss-Prot No. Q9R207]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4683
Target Species Human
Target Species Murine