Please login to your online account to display your discounted pricing

anti-Nbs1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 350-400). [Swiss-Prot No. Q9R207]
Antigen Nbs1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP106609SS

 
novus biologicals canada
NBP106609SS
Unlabeled 0.025mL Each for $139.00

NBP106609

 
novus biologicals canada
NBP106609
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 350-400). [Swiss-Prot No. Q9R207]
Antigen Nbs1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 4683
Formulation PBS, 30% glycerol
Concentration 1mg/mL