missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Nbs1, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 150-200). [Swiss-Prot No. Q9R207]
Antigen Nbs1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP106705 Novus Biologicals Canada
NBP106705
0.1mL Each for $309.05
NBP106705SS Novus Biologicals Canada
NBP106705SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
Nbs1
Affinity Purified
RUO
4683
1.48mg/mL
Synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 150-200). [Swiss-Prot No. Q9R207]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
PBS, 30% glycerol