Please login to your online account to display your discounted pricing

anti-NBR1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human NBR1 protein (within residues 50-250). [Swiss-Prot Q14596]
Antigen NBR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171703

 
novus biologicals canada
NBP171703
Unlabeled 0.1mL Each for $329.00

NBP171703SS

 
novus biologicals canada
NBP171703SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human NBR1 protein (within residues 50-250). [Swiss-Prot Q14596]
Antigen NBR1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Immunogen has 75% identity to the rat protein.
Gene Accession No. Q14596
Gene ID (Entrez) 4077
Formulation PBS
Concentration 1.24mg/mL