missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NANOS1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen This antibody was raised against NANOS1
Antigen NANOS1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227084SS Novus Biologicals Canada
NBP227084SS
0.025mg Each for $109.00
NBP227084 Novus Biologicals Canada
NBP227084
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
NANOS1
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee.
100μg in 200μL PBS containing 0.05% BSA.
This antibody was raised against NANOS1
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
340719
0.5mg/mL