Please login to your online account to display your discounted pricing

anti-Nanog, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse Nanog protein (within residues 1-50). [Swiss-Prot Q80Z64 ]
Antigen Nanog
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP213177

 
novus biologicals canada
NBP213177
Unlabeled 0.1mL Each for $309.05

NBP213177SS

 
novus biologicals canada
NBP213177SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse Nanog protein (within residues 1-50). [Swiss-Prot Q80Z64 ]
Antigen Nanog
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 79923
Formulation PBS, 30% glycerol
Concentration 1.0mg/mL