Please login to your online account to display your discounted pricing

anti-Nanog, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of human NANOG was used as the immunogen.
Antigen Nanog
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224941

 
novus biologicals canada
NBP224941
Unlabeled 0.1mg Each for $329.00

NBP224941SS

 
novus biologicals canada
NBP224941SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of human NANOG was used as the immunogen.
Antigen Nanog
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity This antibody is reactive to Chimpanzee, Goat, Human, Mouse, Rhesus Monkey.
Gene Accession No. Q9H9S0
Gene ID (Entrez) 79923
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL