Please login to your online account to display your discounted pricing

anti-NALP4, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Partial recombinant human NALP4 protein (between residues 1-300) [UniProt Q96MN2]
Antigen NALP4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231375

 
novus biologicals canada
NBP231375
Unlabeled 0.1mg Each for $259.02

NBP231375SS

 
novus biologicals canada
NBP231375SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Partial recombinant human NALP4 protein (between residues 1-300) [UniProt Q96MN2]
Antigen NALP4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Besides detecting canonical isoform (∽113.4 kDa), this antibody may detect isoform 3 (104 kDa) also. Human. Immunogen shows 51% similarity with Mouse's NALP4.
Gene Accession No. Q96MN2
Gene ID (Entrez) 147945
Formulation PBS
Concentration 1.0mg/mL