Please login to your online account to display your discounted pricing

anti-NALP4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056156SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056156T

  • / Each
 

Description & Specifications

Specifications

Antigen NALP4
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 139-157 (FAPKETGKQPRTVIIQGPQ) of human NALP4/PAN2/PYPAF4 was used as immunogen; GenBank no. gi|17064172|gb|AAL35293.1|AF442488_1
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 147945
Test Specificity NALP4 .
Target Species Human
Target Species Murine