Please login to your online account to display your discounted pricing

anti-NAIP Polyclonal, Thermo Scientific™

NAIP Polyclonal Ab PA5-20063 from Thermo Scientific™ specifically detects NAIP in Hu samples, and it is validated for ICC, IF and WB

$506.05 - $506.05

Specifications

Antigen NAIP
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Applications Immunocytochemistry
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PIPA520063

 
pierce biotechnology inc
PA520063
Each for $506.05
Description & Specifications

Specifications

Antigen NAIP
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Quantity 100μg
Regulatory Status RUO
Type Primary
Formulation 100μg purified antibody (1mg/mL) in PBS and 0.02% sodium azide
Gene Symbols NAIP
Immunogen A synthetic peptide corresponding to 13 amino acids at the C-terminus of human NAIP.
Storage Requirements Store at 2-8°C for up to 3 months. For long term storage store at -20°C

NAIP Polyclonal Ab PA5-20063 from Thermo Scientific™ specifically detects NAIP in Hu samples, and it is validated for ICC, IF and WB

Neuronal apoptosis inhibitor protein (NAIP) was the first human inhibitor of apoptosis protein (IAP) identified and was discovered by its association with the neurodegenerative disorder spinal muscular atrophy. Members of the IAP family contain one to three copies of an approximately 70 amino acid motif termed baculovirus IAP repeat (BIR); these BIRs promote protein-protein interactions with various caspases such as caspase-3, -7, and -9 as well as members of the TRAF family of signal molecules (reviewed in 3). Unlike other IAPs however, NAIP requires ATP to bind caspase-9 and is not inhibited by the IAP-inhibiting molecule Smac/DIABLO, suggesting that NAIP is unique among the IAPs in its regulation of its activity. Finally, although only one human NAIP protein has been identified, other shorter NAIP mRNA transcripts have been reported.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: A synthetic peptide corresponding to 13 amino acids at the C-terminus of human NAIP.
Formulation: 100μg purified Antibody (1mg/mL) in PBS, with 0.02% sodium azide;
Synonyms: BIRC1, NLRB1, psiNAIP; baculoviral IAP repeat-containing protein 1; neuronal apoptosis inhibitory protein; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein
Uniprot Accession: Q13075
Gene Symbol: NAIP
Entrez GeneID: 4671
PA5-20063 detects NAIP in Human samples. PA5-20063 has been successfully used in Immunocytochemistry, Immunofluorescence and Western Blot procedures. PA5-20063 The immunogen is A synthetic peptide corresponding to 13 amino acids at the C-terminus of human NAIP..