Please login to your online account to display your discounted pricing

anti-Myosin VB, Polyclonal, Novus Biologicals™

$332.66 - $332.66

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Myosin VB
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

46150002

 
novus biologicals canada
46150002
Unlabeled 0.05mg Each for $332.66
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the Middle Region of the target protein.
Antigen Myosin VB
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity This product is specific for Human MYO5B.
Gene Accession No. Q9ULV0
Gene ID (Entrez) 4645
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0