Please login to your online account to display your discounted pricing

anti-MyoD, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human MyoD1 protein (within residues 1-150). [Swiss-Prot P15172]
Antigen MyoD
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP154153

 
novus biologicals canada
NBP154153
Unlabeled 0.1mL Each for $329.00

NBP154153SS

 
novus biologicals canada
NBP154153SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human MyoD1 protein (within residues 1-150). [Swiss-Prot P15172]
Antigen MyoD
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Immunogen has 90% identity to mouse.
Gene Accession No. P15172
Gene ID (Entrez) 4654
Formulation PBS, 0.1% BSA, 50% glycerol
Concentration 0.58mg/mL