Please login to your online account to display your discounted pricing

anti-MyoD, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP154153SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP154153SS

  • / Each
 

Description & Specifications

Specifications

Antigen MyoD
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 0.58mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS, 0.1% BSA, 50% glycerol
Gene Accession No. P15172
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human MyoD1 protein (within residues 1-150). [Swiss-Prot P15172]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4654
Test Specificity Human. Immunogen has 90% identity to mouse.
Target Species Human