Please login to your online account to display your discounted pricing

anti-MYH, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH.
Antigen MYH
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001032SS

 
novus biologicals canada
NB6001032SS
Unlabeled 0.025mg Each for $139.00

NB6001032

 
novus biologicals canada
NB6001032
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 531-546 of human MYH.
Antigen MYH
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Test Specificity Cross-reacts with Human, Mouse, Rat and Bacteria. Not yet tested in other species.
Gene Accession No. NP_001121897
Gene ID (Entrez) 4595
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL