Please login to your online account to display your discounted pricing

anti-Myeloid-Associated Differentiation Marker, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224494

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224494

  • / Each
 

Description & Specifications

Specifications

Antigen Myeloid-Associated Differentiation Marker
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Gene Accession No. Q96S97
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human MYADM was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 91663
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), horse, rat (isoform CRA_a), mouse (isoforms CRA_b and isoform CRA_d), chimpanzee, dog and rhesus monkey, 93% homologous in opossum and 86% homologous in zebrafish.
Target Species Canine
Target Species Human
Target Species Primate
Target Species Rat