Please login to your online account to display your discounted pricing

anti-Myelin Basic Protein Polyclonal, Thermo Scientific™

Myelin Basic Protein Polyclonal Ab PA1-10008 from Thermo Scientific™ detects Myelin Basic Protein in Bv, Hu, Ms and Rt samples, and is validated for ICC, IF, IHC and WB

$448.05 - $578.55

Specifications

Antigen Myelin Basic Protein
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-265

 
pierce biotechnology inc
PA110008
Unlabeled 100μL Each for $448.05

01-670-572

 
pierce biotechnology inc
PA118319
Unlabeled 100μL Each for $506.05

01-670-577

 
pierce biotechnology inc
PA118324
Unlabeled 100μL Each for $578.55
Description & Specifications

Specifications

Antigen Myelin Basic Protein
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Chicken
Immunogen Full length purified myelin basic protein isolated from cow nerve
Isotype IgY
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Myelin Basic Protein Polyclonal Ab PA1-10008 from Thermo Scientific™ detects Myelin Basic Protein in Bv, Hu, Ms and Rt samples, and is validated for ICC, IF, IHC and WB

Myelin Basic Protein (MBP) is one of the major proteins of the myelin sheath surrounding axons in the nervous system. Since it is of relatively low molecular weight and high abundance the protein sequence was determined from purified protein over 30 years ago. The protein is made by oligodendrocytes in the central nervous system so antibodies to MBP are good markers of this cell type. However, transcripts from the same gene are also expressed in certain hematopoetic lineage cells. In the central nervous system there are three different forms of the protein made by alternate transcription from a single gene, which have molecular weights of 21.5, 18.5 and 17.2 kDa. Since the two lower molecular weight forms are very close in molecular size, MBP antibodies typical show two bands on Western blots, one at about 22kDa and another at about 18 kDa.
Host Species: Chicken
Cross Reactivity: Bovine, Human, Mouse, Rat
Isotype: IgY
Immunogen: Full length purified myelin basic protein isolated from cow nerve
Formulation: 100μL purified Antibody in PBS, with 10mM sodium azide;
Synonyms: MGC99675, MBP
Uniprot Accession: P02686, P02687, P02688, P04370, P15720, P81558, P83487
Gene Symbol: MBP
Entrez GeneID: 100063306, 17196, 24547, 396217, 414286, 4155, 618684
PA1-10008 detects Myelin Basic Protein in Bovine, Human, Mouse and Rat samples. PA1-10008 has been successfully used in Immunocytochemistry, Immunofluorescence, Immunohistochemistry and Western Blot procedures. PA1-10008 The immunogen is Full length purified myelin basic protein isolated from cow nerve.