Please login to your online account to display your discounted pricing

anti-MyD88, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human MyD88 was used as immunogen for this antibody.
Antigen MyD88
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224671

 
novus biologicals canada
NBP224671
Unlabeled 0.1mg Each for $259.02

NBP224671SS

 
novus biologicals canada
NBP224671SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human MyD88 was used as immunogen for this antibody.
Antigen MyD88
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in horse, opossum, chimpanzee, dog, 88% homologous in rat, mouse and 77% homologous in cow.
Gene ID (Entrez) 4615
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL